Please login to your online account to display your discounted pricing

2-(1-Adamantyl)propan-2-ol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
CAS 775-64-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H22O
Formula Weight 194.32
MDL Number MFCD00167855
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

NRB01891CB

 
fisher scientific company
NRB01891CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.32
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C13H22O
CAS 775-64-4
Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00167855