Please login to your online account to display your discounted pricing

2-(1-Adamantyl)propan-2-ol, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  NRB01891CB

 View more versions of this product

Catalog No. NRB01891CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.32
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C13H22O
CAS 775-64-4
Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00167855

Specifications

Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
CAS 775-64-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H22O
Formula Weight 194.32
MDL Number MFCD00167855