Learn More
2-(1-Adamantyl)acetic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB13005DA
Specifications
2-(1-Adamantyl)acetic acid | |
96.5 | |
97% | |
C12H17O2 | |
1 g | |
AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
2-(adamantan-1-yl)acetate | |
123221 | |
97% |
4942-47-6 | |
100.0 | |
Amber Glass Bottle | |
MFCD00074728 | |
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2 | |
193.27 | |
194.27 |
Chemical Identifiers
4942-47-6 | |
193.27 | |
AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
123221 |
C12H17O2 | |
MFCD00074728 | |
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning