missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Adamantyl)-4-methylphenol, 99%

Manufacturer:  Alfa Aesar™ H55922MD

 View more versions of this product

Catalog No. AAH55922MD



Description

Description

Applications
2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Sparingly Soluble in water (7.8E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-(1-Adamantyl)-4-methylphenol
128°C to 130°C
41031-50-9
MFCD00168147
XHLJIHBDAJFXBE-UHFFFAOYSA-N
2-(1-adamantyl)-4-methylphenol
617992
Powder
White
250mg
C17H22O
2-1-adamantyl-4-methyl-phenol
CC1=CC(=C(C=C1)O)C23CC4CC(C2)CC(C4)C3
242.362
242.36
99%

Chemical Identifiers

41031-50-9
242.362
XHLJIHBDAJFXBE-UHFFFAOYSA-N
617992
CC1=CC(=C(C=C1)O)C23CC4CC(C2)CC(C4)C3
C17H22O
MFCD00168147
2-1-adamantyl-4-methyl-phenol
2-(1-adamantyl)-4-methylphenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents