missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[1-(4-Piperonyl)piperazinyl]benzothiazole, Tocris

5-HT4 agonist; Also 5-HT3 antagonist

$168.15 - $697.40

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C19H19N3O2S
Formula Weight 353.44
Chemical Name or Material 2-[1-(4-Piperonyl)piperazinyl]benzothiazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
073650 R & D Systems
RDS073650
50mg Each for $697.40
073610 R & D Systems
RDS073610
10mg Each for $168.15
Specifications Description & Specifications

Chemical Identifiers

>98%
C19H19N3O2S
353.44
2-[1-(4-Piperonyl)piperazinyl]benzothiazole

Specifications

155106-73-3
C19H19N3O2S
50mg
>98%
353.44
2-[1-(4-Piperonyl)piperazinyl]benzothiazole

Safety and Handling

Recommended Storage : Store at Room Temperature