Please login to your online account to display your discounted pricing

2-(1,4-Diazepan-1-yl)nicotinonitrile, ≥95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00102CB

 View more versions of this product

Catalog No. MO00102CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.26
Quantity 250mg
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
CAS 352018-97-4
Chemical Name or Material 2-(1, 4-Diazepan-1-yl)nicotinonitrile
Molecular Formula C11H14N4
MDL Number MFCD03086210
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-(1, 4-Diazepan-1-yl)nicotinonitrile
CAS 352018-97-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H14N4
Formula Weight 202.26
MDL Number MFCD03086210
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.