Please login to your online account to display your discounted pricing

2-(1,4-Diazepan-1-yl)nicotinonitrile, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(1, 4-Diazepan-1-yl)nicotinonitrile
CAS 352018-97-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H14N4
Formula Weight 202.26
MDL Number MFCD03086210
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00102CB

  SDS
fisher scientific company
MO00102CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO00102DA

  SDS
fisher scientific company
MO00102DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.26
Quantity 250mg
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
CAS 352018-97-4
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
MDL Number MFCD03086210
Packaging Amber Glass Bottle
Molecular Formula C11H14N4
Chemical Name or Material 2-(1, 4-Diazepan-1-yl)nicotinonitrile
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.