Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1,3-Dioxolan-2-on-4-yl)-1-ethylboronic acid pinacol ester, 97%

$236.14 - $236.14

Chemical Identifiers

Chemical Name or Material 2-(1,3-Dioxolan-2-on-4-yl)-1-ethylboronic acid pinacol ester
CAS 501014-47-7
Molecular Formula C11H19BO5
Formula Weight 242.08g/mol
MDL Number MFCD03788724
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1953303

 
alfa aesar™
L19533-03
1g Each for $236.14
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-(1,3-Dioxolan-2-on-4-yl)-1-ethylboronic acid pinacol ester
CAS 501014-47-7
Assay 97%
Molecular Formula C11H19BO5
Formula Weight 242.08g/mol
MDL Number MFCD03788724