Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester
Molecular Formula C17H21BN2O5S
Formula Weight 376.23
MDL Number MFCD16294525
Synonym 1,3-Dimethyl-5-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienylmethylene]pyrimidine-2,4,6-trione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5189806

  SDS
alfa aesar™
H5189806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5189803

  SDS
alfa aesar™
H5189803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 376.23
MDL Number MFCD16294525
Molecular Formula C17H21BN2O5S
Synonym 1,3-Dimethyl-5-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienylmethylene]pyrimidine-2,4,6-trione
Assay 95%
Quantity 5g
Chemical Name or Material 2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester