Please login to your online account to display your discounted pricing

2,1,3-Benzoxadiazol-4-ylmethanol, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC09609DA

 View more versions of this product

Catalog No. CC09609DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 150.14
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-ylmethanol
Assay Percent Range 97%
Molecular Formula C7H6N2O2
MDL Number MFCD04115376
CAS 175609-19-5
Packaging Amber glass bottle
Synonym 2, 1, 3-Benzoxadiazol-4-ylmethanol (IUPAC)
CAS Min % 96.5

Specifications

Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-ylmethanol
CAS 175609-19-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H6N2O2
Formula Weight 150.14
MDL Number MFCD04115376
Synonym 2, 1, 3-Benzoxadiazol-4-ylmethanol (IUPAC)