Please login to your online account to display your discounted pricing

2-(1,3-Benzothiazol-2-yl)acetonitrile, 97%, Maybridge


Quantity 25g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  GK00309EB

 View more versions of this product

Catalog No. GK00309EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 174.23
Quantity 25g
Assay 97%
Assay Percent Range 97%
CAS 56278-50-3
Chemical Name or Material 2-(1, 3-Benzothiazol-2-yl)acetonitrile
Molecular Formula C9H6N2S
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-(1, 3-Benzothiazol-2-yl)acetonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6N2S
Formula Weight 174.23