missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1,3-Benzothiazol-2-yl)acetonitrile, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H6N2S
Formula Weight 174.23
Chemical Name or Material 2-(1, 3-Benzothiazol-2-yl)acetonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
GK00309EB Fisher Scientific Company
GK00309EB
Amber Glass Bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
174.23
97%
C9H6N2S
2-(1, 3-Benzothiazol-2-yl)acetonitrile

Specifications

174.23
97%
C9H6N2S
2-(1, 3-Benzothiazol-2-yl)acetonitrile
25g
56278-50-3
Amber Glass Bottle
97%