missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,1,3-Benzothiadiazole, 98%

Manufacturer:  Alfa Aesar™ 4373009

 View more versions of this product

Catalog No. AA4373009



Specifications

Specifications

2,1,3-Benzothiadiazole
206°C
10g
C6H4N2S
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
C1=CC2=NSN=C2C=C1
136.172
136.18
98%
95°C (203°F)
42°C to 44°C
273-13-2
MFCD00005809
PDQRQJVPEFGVRK-UHFFFAOYSA-N
2,1,3-benzothiadiazole
67502
Powder

Chemical Identifiers

273-13-2
136.172
PDQRQJVPEFGVRK-UHFFFAOYSA-N
67502
C1=CC2=NSN=C2C=C1
C6H4N2S
MFCD00005809
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
2,1,3-benzothiadiazole
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents