Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC09201DA

 View more versions of this product

Catalog No. CC09201DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C7H4N2O2S
MDL Number MFCD01647555
CAS 16405-98-4
CAS Min % 96.5

Specifications

Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carboxylic acid
CAS 16405-98-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4N2O2S
Formula Weight 180.19
MDL Number MFCD01647555