Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC09204DA

 View more versions of this product

Catalog No. CC09204DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 164.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carbaldehyde
CAS Min % 96.5
MDL Number MFCD00457949
Assay Percent Range 97%
Packaging Amber glass bottle
CAS 71605-72-6
Molecular Formula C7H4N2OS

Specifications

Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carbaldehyde
CAS 71605-72-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4N2OS
Formula Weight 164.19
MDL Number MFCD00457949