missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC09103DE

 View more versions of this product

Catalog No. CC09103DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 1, 3-Benzothiadiazole-4-sulfonyl chloride
96.5
97%
MFCD00068049
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
ClS(=O)(=O)C1=CC=CC2=NSN=C12
234.67
234.69
73713-79-8
100.0
C6H3ClN2O2S2
5g
CXAICGCTHOWKPP-UHFFFAOYSA-N
2,1,3-benzothiadiazole-4-sulfonyl chloride
2735459
97%

Chemical Identifiers

73713-79-8
234.67
CXAICGCTHOWKPP-UHFFFAOYSA-N
2735459
ClS(=O)(=O)C1=CC=CC2=NSN=C12
C6H3ClN2O2S2
MFCD00068049
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
2,1,3-benzothiadiazole-4-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions