Learn More
2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™
$318.78 - $1385.66
Chemical Identifiers
CAS | 73713-79-8 |
---|---|
Molecular Formula | C6H3ClN2O2S2 |
Molecular Weight (g/mol) | 234.67 |
MDL Number | MFCD00068049 |
InChI Key | CXAICGCTHOWKPP-UHFFFAOYSA-N |
Synonym | benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride |
PubChem CID | 2735459 |
IUPAC Name | 2,1,3-benzothiadiazole-4-sulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CC=CC2=NSN=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5037703
|
Thermo Scientific Chemicals
H5037703 |
1 g |
Each for $318.78
|
|
|||||
AAH5037706
|
Thermo Scientific Chemicals
H5037706 |
5 g |
Each for $1,385.66
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
73713-79-8 | |
234.67 | |
CXAICGCTHOWKPP-UHFFFAOYSA-N | |
2735459 | |
ClS(=O)(=O)C1=CC=CC2=NSN=C12 |
C6H3ClN2O2S2 | |
MFCD00068049 | |
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride | |
2,1,3-benzothiadiazole-4-sulfonyl chloride |
Specifications
73713-79-8 | |
C6H3ClN2O2S2 | |
1 g | |
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride | |
ClS(=O)(=O)C1=CC=CC2=NSN=C12 | |
234.67 | |
234.68 | |
2,1,3-Benzothiadiazole-4-sulfonyl chloride |
145°C to 147°C | |
MFCD00068049 | |
UN3261 | |
CXAICGCTHOWKPP-UHFFFAOYSA-N | |
2,1,3-benzothiadiazole-4-sulfonyl chloride | |
2735459 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber : DL0100000
TSCA : No
Recommended Storage : Ambient temperatures