Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC09104DA

 View more versions of this product

Catalog No. CC09104DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 164.19
Quantity 1g
Assay 95%
CAS Max % 100.0
CAS 5170-68-3
Assay Percent Range 95%
MDL Number MFCD02681902
CAS Min % 94.5
Molecular Formula C7H4N2OS
Packaging Amber Glass Bottle
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carbaldehyde

Specifications

Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carbaldehyde
CAS 5170-68-3
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H4N2OS
Formula Weight 164.19
MDL Number MFCD02681902