missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazol-4-yl isocyanate, 97%, Maybridge

Chemical Identifiers

CAS 342411-14-7
Molecular Formula C7H3N3OS
Molecular Weight (g/mol) 177.181
MDL Number MFCD02681903
InChI Key HPZZFYLZFYDNMA-UHFFFAOYSA-N
Synonym 2,1,3-benzothiadiazol-4-yl isocyanate
PubChem CID 2776284
IUPAC Name 4-isocyanato-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C(=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC09106CB SDS View Documents Fisher Scientific Company
CC09106CB
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC09106DE SDS View Documents Fisher Scientific Company
CC09106DE
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

342411-14-7
177.181
HPZZFYLZFYDNMA-UHFFFAOYSA-N
2776284
C1=CC2=NSN=C2C(=C1)N=C=O
C7H3N3OS
MFCD02681903
2,1,3-benzothiadiazol-4-yl isocyanate
4-isocyanato-2,1,3-benzothiadiazole

Specifications

Amber Glass Bottle
342411-14-7
100.0
C7H3N3OS
2,1,3-benzothiadiazol-4-yl isocyanate
C1=CC2=NSN=C2C(=C1)N=C=O
177.181
177.19
2, 1, 3-Benzothiadiazol-4-yl isocyanate
250mg
96.5
97%
MFCD02681903
HPZZFYLZFYDNMA-UHFFFAOYSA-N
4-isocyanato-2,1,3-benzothiadiazole
2776284
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning