missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzothiadiazol-4-yl isocyanate, 97%, Maybridge

Manufacturer:  Thermo Scientific CC09106DE

 View more versions of this product

Catalog No. CC09106DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 1, 3-Benzothiadiazol-4-yl isocyanate
342411-14-7
100.0
C7H3N3OS
2,1,3-benzothiadiazol-4-yl isocyanate, 2,1,3-benzothiadiazol-4-yl isoxyanate, 4-isocyanatobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazol-4-ylisocyanate, 2,1,3-benzothiadiazole,4-isocyanato, benzo c 1,2,5-thiadiazol-4-isocyanate, 2,1,3-benzothiadiazole, 4-isocyanato, 2,1,3-benzothiadiazole, 4-isocyanato-9ci
C1=CC2=NSN=C2C(=C1)N=C=O
177.181
177.19
5g
96.5
97%
MFCD02681903
HPZZFYLZFYDNMA-UHFFFAOYSA-N
4-isocyanato-2,1,3-benzothiadiazole
2776284
97%

Chemical Identifiers

342411-14-7
177.181
HPZZFYLZFYDNMA-UHFFFAOYSA-N
2776284
C1=CC2=NSN=C2C(=C1)N=C=O
C7H3N3OS
MFCD02681903
2,1,3-benzothiadiazol-4-yl isocyanate, 2,1,3-benzothiadiazol-4-yl isoxyanate, 4-isocyanatobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazol-4-ylisocyanate, 2,1,3-benzothiadiazole,4-isocyanato, benzo c 1,2,5-thiadiazol-4-isocyanate, 2,1,3-benzothiadiazole, 4-isocyanato, 2,1,3-benzothiadiazole, 4-isocyanato-9ci
4-isocyanato-2,1,3-benzothiadiazole
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents