missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,1,3-Benzoselenadiazole, TCI America™

$278.06 - $960.98

Chemical Identifiers

CAS 273-15-4
Molecular Formula C6H4N2Se
Molecular Weight (g/mol) 183.083
MDL Number MFCD00022886
InChI Key AYTPIVIDHMVGSX-UHFFFAOYSA-N
PubChem CID 67503
IUPAC Name 2,1,3-benzoselenadiazole
SMILES C1=CC2=N[Se]N=C2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4313200MG View Documents TCI America
B4313200MG
200mg Each for $278.06
 
B43131G View Documents TCI America
B43131G
1g Each for $960.98
 
Specifications

Chemical Identifiers

273-15-4
183.083
AYTPIVIDHMVGSX-UHFFFAOYSA-N
2,1,3-benzoselenadiazole
C6H4N2Se
MFCD00022886
67503
C1=CC2=N[Se]N=C2C=C1

Specifications

246°C
75°C
273-15-4
MFCD00022886
AYTPIVIDHMVGSX-UHFFFAOYSA-N
2,1,3-benzoselenadiazole
67503
Crystal-Powder at 20°C
2,1,3-Benzoselenadiazole
Undesignated
200mg
C6H4N2Se
3283
C1=CC2=N[Se]N=C2C=C1
183.083
183.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No