2-(1,1-Difluoroethyl)pyrazine, 97%, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC451360010
 View more versions of this product
Catalog No. AC451360010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
2-(1, 1-Difluoroethyl)pyrazine
150.0°C to 152.0°C
111781-49-8
C6H6F2N2
OXKFWGSACYGCSY-UHFFFAOYSA-N
2-(1,1-difluoroethyl)pyrazine
19763564
97%
53°C
1g
96% min. (GC)
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl
CC(C1=NC=CN=C1)(F)F
144.12
144.12
Chemical Identifiers
111781-49-8
144.12
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl
2-(1,1-difluoroethyl)pyrazine
C6H6F2N2
OXKFWGSACYGCSY-UHFFFAOYSA-N
19763564
CC(C1=NC=CN=C1)(F)F
Safety and Handling
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS Signal Word: Warning