missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(1,1-Difluoroethyl)pyrazine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451360050
Specifications
2-(1, 1-Difluoroethyl)pyrazine | |
150.0°C to 152.0°C | |
111781-49-8 | |
C6H6F2N2 | |
OXKFWGSACYGCSY-UHFFFAOYSA-N | |
2-(1,1-difluoroethyl)pyrazine | |
19763564 | |
97% |
53°C | |
5g | |
96% min. (GC) | |
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl | |
CC(C1=NC=CN=C1)(F)F | |
144.12 | |
144.12 |
Chemical Identifiers
111781-49-8 | |
144.12 | |
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl | |
2-(1,1-difluoroethyl)pyrazine |
C6H6F2N2 | |
OXKFWGSACYGCSY-UHFFFAOYSA-N | |
19763564 | |
CC(C1=NC=CN=C1)(F)F |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS Signal Word: Warning