Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S)-(+)-Menthyl glyoxylate monohydrate, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1978506

 View more versions of this product

Catalog No. AAL1978506

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 230.31 (212.29 Anhydrous)
Chemical Name or Material (1S)-(+)-Menthyl glyoxylate monohydrate
MDL Number MFCD04037548
Health Hazard 2 GHS H Statement
H318-H373
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P261-P280d-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing and eye protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 98%
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
UN Number UN3077
Molecular Formula C12H22O4
Quantity 5g
Melting Point 76° to 80°C
Beilstein 4905777
Health Hazard 1 Danger
Synonym Glyoxylic acid (1S)-menthyl ester hydrate
Optical Rotation +74° (c=1 in Methanol)
Boiling Point 95° to 96°C (0.3mmHg)

Specifications

Chemical Name or Material (1S)-(+)-Menthyl glyoxylate monohydrate
Molecular Formula C12H22O4
Formula Weight 230.31 (212.29 Anhydrous)
MDL Number MFCD04037548
UN Number UN3077
Beilstein 4905777
Synonym Glyoxylic acid (1S)-menthyl ester hydrate
Accessories

Danger

GHS P Statement
P261-P280d-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing and eye protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H318-H373
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents