Please login to your online account to display your discounted pricing

(1S)-(+)-Menthyl chloroformate, 97%, ACROS Organics™

$90.23 - $90.23

Chemical Identifiers

Chemical Name or Material (1S)-(+)-Menthyl chloroformate
Name Note 97%
CAS 7635-54-3
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C11H19ClO2
Formula Weight 218.73g/mol
MDL Number MFCD00134483
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314510250

  SDS
thermo fisher scientific
314510250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC314510050

  SDS
thermo fisher scientific
314510050
Glass bottle 5mL Each for $90.23
Description & Specifications

Specifications

Density 1.02g/mL
Infrared Spectrum Authentic
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Packaging Glass bottle
Specific Rotation +78° to +88° (20°C, 589nm) (c=1, CHCl3)
Specific Rotation Condition +83° (20°C c=1,CHCl3)
Color Clear colorless to brown
Quantity 25mL
Chemical Name or Material (1S)-(+)-Menthyl chloroformate
Name Note 97%
CAS 7635-54-3
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear colorless to brown liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C11H19ClO2
Formula Weight 218.73g/mol
MDL Number MFCD00134483
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if inhaled.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

Coupling reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if inhaled.
Contact with water liberates toxic gas.