Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S)-(+)-Camphor-10-sulfonyl chloride, 97%

$12.93 - $12.93

Chemical Identifiers

Chemical Name or Material (1S)-(+)-Camphor-10-sulfonyl chloride
CAS 21286-54-4
Molecular Formula C10H15ClO3S
Formula Weight 250.74
MDL Number MFCD00064156
UN Number UN3261
Beilstein 3205974
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1329306

  SDS
alfa aesar™
A1329306
5g Each for $12.93

AAA1329314

  SDS
alfa aesar™
A1329314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1329322

  SDS
alfa aesar™
A1329322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.74
Chemical Name or Material (1S)-(+)-Camphor-10-sulfonyl chloride
Assay 97%
Molecular Formula C10H15ClO3S
MDL Number MFCD00064156
CAS 21286-54-4
UN Number UN3261
Beilstein 3205974
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 64° to 68°C
Quantity 5g
Optical Rotation +32° (c=1 in Chloroform)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.