Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S)-(-)-Camphanic chloride, 95%

Chemical Identifiers

Chemical Name or Material (1S)-(-)-Camphanic chloride
CAS 39637-74-6
Molecular Formula C10H13ClO3
Formula Weight 216.67
MDL Number MFCD00135626
UN Number UN3261
Beilstein 3590860
Synonym (-)-Camphanic acid chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1414803

  SDS
alfa aesar™
L1414803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1414806

  SDS
alfa aesar™
L1414806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.67
CAS 39637-74-6
Chemical Name or Material (1S)-(-)-Camphanic chloride
Assay 95%
Molecular Formula C10H13ClO3
MDL Number MFCD00135626
UN Number UN3261
Synonym (-)-Camphanic acid chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Beilstein 3590860
Melting Point 64° to 68°C
Quantity 1g
Optical Rotation −18° (c=2 in Carbon Tetrachloride)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
(1S)-(-)-Camphanic chloride is used as a resolving agent for alcohols as the diastereomeric esters by crystallization or chromatography and as a chiral derivatization reagent for determination of enantiomeric excess of alcohols and amines.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to moisture. Protect from humidity and water. Incompatible with bases and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

−30° to −10°C