Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S)-(-)-Camphanic acid, 99%

$62.96 - $201.65

Chemical Identifiers

Chemical Name or Material (1S)-(-)-Camphanic acid
CAS 13429-83-9
Molecular Formula C10H14O4
Formula Weight 198.22
MDL Number MFCD00044948
Beilstein 84570
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1609803

 
alfa aesar™
L1609803
1g Each for $62.96

AAL1609806

 
alfa aesar™
L1609806
5g Each for $201.65
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material (1S)-(-)-Camphanic acid
MDL Number MFCD00044948
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Molecular Formula C10H14O4
CAS 13429-83-9
Beilstein 84570
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 198° to 202°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.