Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 98%

$82.97 - $784.81

Chemical Identifiers

Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
CAS 113451-59-5
Molecular Formula C10H18N2O2
Formula Weight 198.26g/mol
MDL Number MFCD01569250
Synonym tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5109703

 
alfa aesar™
H51097-03
Each for $198.51

AAH5109706

 
alfa aesar™
H51097-06
Each for $784.81

AAH51097MD

 
alfa aesar™
H51097-MD
Each for $82.97
Description & Specifications

Specifications

Melting Point 74° to 76°C
Quantity 1g
Optical Rotation −44° (c=1 in Chloroform)
Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
CAS 113451-59-5
Assay 98%
Molecular Formula C10H18N2O2
Formula Weight 198.26g/mol
MDL Number MFCD01569250
Synonym tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C