missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 398040010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane | |
94.0 | |
Authentic | |
Glass bottle | |
MFCD01569250 | |
-41° to -47° (20°C, 589nm) (C=1, chloroform) | |
Solubility in water: insoluble | |
CC(C)(C)OC(=O)N1CC2CC1CN2 | |
198.26 | |
198.26 |
113451-59-5 | |
100.0 | |
94% min. (GC) | |
C10H18N2O2 | |
1g | |
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate | |
UXAWXZDXVOYLII-YUMQZZPRSA-N | |
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate | |
11521263 | |
95% |
Chemical Identifiers
113451-59-5 | |
198.26 | |
UXAWXZDXVOYLII-YUMQZZPRSA-N | |
11521263 | |
CC(C)(C)OC(=O)N1CC2CC1CN2 |
C10H18N2O2 | |
MFCD01569250 | |
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate | |
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only