missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide, TCI America™

$178.75 - $690.56

Chemical Identifiers

CAS 116258-17-4
Molecular Formula C12H18Br2N2
Molecular Weight (g/mol) 350.098
MDL Number MFCD01321292
InChI Key SOMPEQIPSQFVMO-AQEKLAMFSA-N
PubChem CID 14612542
IUPAC Name (1S,4S)-2-benzyl-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
SMILES C1C2CNC1CN2CC3=CC=CC=C3.Br.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20541G View Documents TCI America
B20541G
1g Each for $178.75
 
B20545G View Documents TCI America
B20545G
5g Each for $690.56
 
Specifications

Chemical Identifiers

116258-17-4
350.098
SOMPEQIPSQFVMO-AQEKLAMFSA-N
(1S,4S)-2-benzyl-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
C12H18Br2N2
MFCD01321292
14612542
C1C2CNC1CN2CC3=CC=CC=C3.Br.Br

Specifications

White
116258-17-4
MFCD01321292
C1C2CNC1CN2CC3=CC=CC=C3.Br.Br
350.098
350.10
≥98.0% (T)
1g
C12H18Br2N2
SOMPEQIPSQFVMO-AQEKLAMFSA-N
(1S,4S)-2-benzyl-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
14612542
Crystal-Powder at 20°C
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide
Safety and Handling

Safety and Handling

TSCA : No