Please login to your online account to display your discounted pricing

(1S,4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid
Name Note 98% ee
CAS 220497-64-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C21 H19 N O4
Formula Weight 349.39
MDL Number MFCD01311756
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361520010

  SDS
thermo fisher scientific
361520010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC361525000

  SDS
thermo fisher scientific
361525000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 349.39
Quantity 1g
Color White
CAS Min % 94.0
CAS Max % 100.0
CAS 220497-64-3
Infrared Spectrum Authentic
Chemical Name or Material (1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid
Assay Percent Range 95%
Molecular Formula C21 H19 N O4
MDL Number MFCD01311756
Name Note 98% ee
Enantiomeric Excess 97.5% min.
Packaging Glass bottle