Please login to your online account to display your discounted pricing

(1S,4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361525000

 View more versions of this product

Catalog No. AC361525000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 349.39
Physical Form Powder
Quantity 500mg
Color White
CAS Min % 94.0
CAS Max % 100.0
CAS 220497-64-3
MDL Number MFCD01311756
Infrared Spectrum Authentic
Chemical Name or Material (1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Molecular Formula C21 H19 N O4
Name Note 98% ee
Assay Percent Range 95%

Specifications

Chemical Name or Material (1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid
Name Note 98% ee
CAS 220497-64-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C21 H19 N O4
Formula Weight 349.39
MDL Number MFCD01311756