missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 361520010

 View more versions of this product

Catalog No. AC361520010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
349.39
White
100.0
220497-64-3
C21 H19 N O4
MFCD01311756
Glass bottle
Powder
1g
94.0
Authentic
95%
(1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid
97.5% min.
98% ee

Chemical Identifiers

220497-64-3
100.0
C21 H19 N O4
MFCD01311756
≥94%
94.0
95%
349.39
(1S, 4R)-N-FMOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 98% ee