missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 361820010
Specifications
(1S, 4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid | |
151907-80-1 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00211288 | |
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
WOUNTSATDZJBLP-UHFFFAOYNA-N | |
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid | |
7019281 | |
95%,98% ee |
98% ee | |
94.0 | |
97.5% min. (HPLC) | |
94% min. (HPLC) | |
C11H17NO4 | |
1g | |
Solubility in water: soluble | |
CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O | |
227.26 | |
227.26 |
Chemical Identifiers
151907-80-1 | |
227.26 | |
WOUNTSATDZJBLP-UHFFFAOYNA-N | |
7019281 | |
CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O |
C11H17NO4 | |
MFCD00211288 | |
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO â Research Use Only