Please login to your online account to display your discounted pricing

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  362220010

 View more versions of this product

Catalog No. AC362220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (1S, 4R)-2-Azabicyclo[2.2.1]heptan-3-one
CAS 134003-03-5
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Physical Form Crystalline Powder
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C6H9NO
Packaging Glass bottle
Formula Weight 111.14
Color White to slightly yellow
Quantity 1g

Specifications

Chemical Name or Material (1S, 4R)-2-Azabicyclo[2.2.1]heptan-3-one
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C6H9NO
Formula Weight 111.14