Please login to your online account to display your discounted pricing

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC362220050

 View more versions of this product

Catalog No. AC362220050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥94% (GC)
Formula Weight 111.14
Quantity 5g
Color White to slightly yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
CAS 134003-03-5
Chemical Name or Material (1S, 4R)-2-Azabicyclo[2.2.1]heptan-3-one
Molecular Formula C6H9NO

Specifications

Chemical Name or Material (1S, 4R)-2-Azabicyclo[2.2.1]heptan-3-one
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C6H9NO
Formula Weight 111.14