missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, ACROS Organics™
Chemical Identifiers
CAS | 161660-94-2 |
---|---|
Molecular Formula | C11H19NO4 |
Molecular Weight (g/mol) | 229.27 |
MDL Number | MFCD01320858 |
InChI Key | RNJQBGXOSAQQDG-SFYZADRCSA-N |
Synonym | --1r,3s-n-boc-3-aminocyclopentane carboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid |
PubChem CID | 1512526 |
IUPAC Name | (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
SMILES | CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC361842500
|
Thermo Fisher Scientific
361842500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC361840010
|
Thermo Fisher Scientific
361840010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
161660-94-2 | |
229.27 | |
RNJQBGXOSAQQDG-SFYZADRCSA-N | |
1512526 | |
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O |
C11H19NO4 | |
MFCD01320858 | |
--1r,3s-n-boc-3-aminocyclopentane carboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid | |
(1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
Specifications
97.5% min. | |
Glass bottle | |
96.1°C | |
161660-94-2 | |
100.0 | |
C11H19NO4 | |
--1r,3s-n-boc-3-aminocyclopentane carboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid | |
RNJQBGXOSAQQDG-SFYZADRCSA-N | |
(1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid | |
1512526 | |
Powder | |
(1S, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.) |
Authentic | |
White | |
250mg | |
94.0 | |
94% min. (HPLC) | |
MFCD01320858 | |
Solubility in water: soluble | |
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O | |
229.27 | |
229.27 | |
95%,98% ee |
Safety and Handling
TSCA : TSCA