missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,3R)-(-)-Camphoric acid, 98%

Manufacturer:  Alfa Aesar™ H54042MD

 View more versions of this product

Catalog No. AAH54042MD



Specifications

Specifications

(1S,3R)-(-)-Camphoric acid
188°C to 190°C
560-09-8
MFCD00064937
14,1733
LSPHULWDVZXLIL-QUBYGPBYSA-N
(1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
219463
98%
−48° (c=10 in Ethanol)
250mg
C10H16O4
1874674
--camphoric acid
CC1(C(CCC1(C)C(=O)O)C(=O)O)C
200.234
200.23

Chemical Identifiers

560-09-8
200.234
LSPHULWDVZXLIL-QUBYGPBYSA-N
219463
CC1(C(CCC1(C)C(=O)O)C(=O)O)C
C10H16O4
MFCD00064937
--camphoric acid
(1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents