Please login to your online account to display your discounted pricing

(1S,3R)-ACPD, Tocris


Group I/group II mGlu agonist

Quantity 10mg

Manufacturer: r & d systems  RDS028410

 View more versions of this product

Catalog No. 028410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 173.17
Chemical Name or Material (1S, 3R)-ACPD
Assay Percent Range >98%
Molecular Formula C7H11NO4
CAS 111900-32-4
Synonym (1S, 3R)-1-Aminocyclopentane-1, 3-dicarboxylic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (1S, 3R)-ACPD
Assay Percent Range >98%
Molecular Formula C7H11NO4
Formula Weight 173.17
Synonym (1S, 3R)-1-Aminocyclopentane-1, 3-dicarboxylic acid
Accessories

Store at Room Temperature