Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid, 95%

$190.86 - $604.93

Chemical Identifiers

Chemical Name or Material (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid
CAS 261165-05-3
Molecular Formula C11H19NO4
Formula Weight 229.28
MDL Number MFCD01320857
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5219403

 
alfa aesar™
H5219403
1g Each for $604.93

AAH52194MD

 
alfa aesar™
H52194MD
250mg Each for $190.86
Description & Specifications

Specifications

Formula Weight 229.28
Chemical Name or Material (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid
Assay 95%
Molecular Formula C11H19NO4
MDL Number MFCD01320857
CAS 261165-05-3
Melting Point 110° to 114°C
Quantity 1g
Optical Rotation +16.0° (c=1 in Methanol)