Please login to your online account to display your discounted pricing

(1S,2S)-trans-1,2-Cyclohexanediol, 98%, Acros Organics


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  337672500

 View more versions of this product

Catalog No. AC337672500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals
Formula Weight 116.16
Quantity 250mg
Color White to off-white
TSCA TSCA
Melting Point 105° to 109°C
CAS Max % 100.0
Chemical Name or Material (1S, 2S)-trans-1, 2-Cyclohexanediol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H12O2
MDL Number MFCD00066431
CAS 57794-08-8
Specific Rotation +39° to +44° (25°C, 589nm) (c=1.5, H2O)
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H10(OH)2

Specifications

Chemical Name or Material (1S, 2S)-trans-1, 2-Cyclohexanediol
Name Note 98%
CAS 57794-08-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O2
Linear Formula C6H10(OH)2
Formula Weight 116.16
MDL Number MFCD00066431