Please login to your online account to display your discounted pricing

(1S,2S)-trans-1,2-Cyclohexanediol, 98%, Acros Organics

$111.70 - $111.70

Chemical Identifiers

Chemical Name or Material (1S, 2S)-trans-1, 2-Cyclohexanediol
Name Note 98%
CAS 57794-08-8
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H12O2
Linear Formula C6H10(OH)2
Formula Weight 116.16g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC337672500

  SDS
thermo fisher scientific
337672500
Glass bottle 250mg Each for $111.70
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +39° to +44° (25°C, 589nm) (c=1.5, H2O)
Color White to off-white
Melting Point 105° to 109°C
Quantity 250mg
Chemical Name or Material (1S, 2S)-trans-1, 2-Cyclohexanediol
Name Note 98%
CAS 57794-08-8
CAS Min % 97.5
CAS Max % 100.0
Physical Form White to off-white crystals
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H12O2
Linear Formula C6H10(OH)2
Formula Weight 116.16g/mol
MDL Number MFCD00066431
TSCA TSCA