missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%

Manufacturer:  Alfa Aesar™ H63445MC

 View more versions of this product

Catalog No. AAH63445MC



Description

Description

Applications
It is employed as a intermediate for pharmaceutical and asymmetirc synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
Air Sensitive
100mg
C44H44N2P2
1s,2s-n1,n1-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n1,n1-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine, 1s,2s-n,n'-bis 2-diphenylphosphino phenyl methyl-1,2-cyclohexanediamine, 1s,2s-n~1~,n~1~-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine
C1CCC(C(C1)N)N(CC2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)CC5=CC=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
662.798
662.78
97%
Unpleasant
Yellow
174677-83-9
MFCD17014022
GEZITKLHMKZYMH-COCZKOEFSA-N
(1S,2S)-2-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
71463644
Solid

Chemical Identifiers

174677-83-9
662.798
GEZITKLHMKZYMH-COCZKOEFSA-N
71463644
C1CCC(C(C1)N)N(CC2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)CC5=CC=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
C44H44N2P2
MFCD17014022
1s,2s-n1,n1-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n1,n1-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine, 1s,2s-n,n'-bis 2-diphenylphosphino phenyl methyl-1,2-cyclohexanediamine, 1s,2s-n~1~,n~1~-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine
(1S,2S)-2-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
SDS Documents