Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%

Chemical Identifiers

Chemical Name or Material (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
CAS 174677-83-9
Formula Weight 662.78
MDL Number MFCD17014022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63445MC

  SDS
alfa aesar™
H63445MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63445ME

  SDS
alfa aesar™
H63445ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 662.78
Chemical Name or Material (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
Odor Unpleasant
Sensitivity Air Sensitive
Physical Form Solid
Assay 97%
MDL Number MFCD17014022
CAS 174677-83-9
Color Yellow
Quantity 100mg

Applications
It is employed as a intermediate for pharmaceutical and asymmetirc synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.