Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine, 98%

$98.89 - $98.89

Chemical Identifiers

Chemical Name or Material (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine
CAS 58520-04-0
Molecular Formula C16H20N2O2
Formula Weight 272.34g/mol
MDL Number MFCD08460191
Synonym (1S,2S)-Bis(2-methoxyphenyl)-1,2-diaminoethane; (S,S)-DAEN
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2580103

 
alfa aesar™
H25801-03
1g Each for $98.89
Description & Specifications

Specifications

Melting Point 89° to 91°C
Quantity 1g
Optical Rotation −77° (c=2 in Chloroform)
Chemical Name or Material (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine
CAS 58520-04-0
Assay 98%
Molecular Formula C16H20N2O2
Formula Weight 272.34g/mol
MDL Number MFCD08460191
Synonym (1S,2S)-Bis(2-methoxyphenyl)-1,2-diaminoethane; (S,S)-DAEN
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.