Please login to your online account to display your discounted pricing

(1S,2S,4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  362322500

 View more versions of this product

Catalog No. AC362322500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 259.3
Physical Form Crystalline Powder
Quantity 250mg
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (1S, 2S, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Packaging Glass bottle
Molecular Formula C12H21NO5
Assay Percent Range 95%
CAS 262280-14-8
MDL Number MFCD02259721
Name Note 95%, (98% e.e.)
Infrared Spectrum Authentic
Synonym Methyl (1R, 3S, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate
Enantiomeric Excess 97.5% min. (GC)

Specifications

Chemical Name or Material (1S, 2S, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
CAS 262280-14-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02259721
Synonym Methyl (1R, 3S, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate