missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2S,3R,5S)-(+)-2,3-Pinanediol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC279551000

 View more versions of this product

Catalog No. AC279551000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1S,2S,3R,5S)-(+)-2,3-Pinanediol
Authentic
+8.50 (20.00°C c=6.5,toluene)
White to Beige
100g
97.5
97.5% min. (GC)
MFCD00077851
+-2-hydroxyisopinocampheol, +-pinanediol, 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-+-pinanediol, 1s,2s,3r,5s-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s, pinanediol, +, unii-y6zcv4avra, y6zcv4avra
CC1(C2CC1C(C(C2)O)(C)O)C
170.25
170.25
98%
>110°C
Glass bottle
+ 8.5
52°C to 60°C
18680-27-8
100.0
C10H18O2
06,4145
MOILFCKRQFQVFS-OORONAJNSA-N
(1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
10219606
Crystalline Powder, Crystals or Mass

Chemical Identifiers

18680-27-8
170.25
MOILFCKRQFQVFS-OORONAJNSA-N
10219606
CC1(C2CC1C(C(C2)O)(C)O)C
C10H18O2
MFCD00077851
+-2-hydroxyisopinocampheol, +-pinanediol, 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-+-pinanediol, 1s,2s,3r,5s-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s, pinanediol, +, unii-y6zcv4avra, y6zcv4avra
(1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SDS Documents