Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Methoxycyclohexanol, ChiPros 99%, ee 98%


Quantity 1g

Manufacturer: alfa aesar™  L1906503

 View more versions of this product

Catalog No. AAL1906503

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 130.19
Chemical Name or Material (1S,2S)-(+)-2-Methoxycyclohexanol
Assay 99%
Molecular Formula C7H14O2
MDL Number MFCD00211253
CAS 134108-92-2
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay Percent Notes ee 98%
Beilstein 1848990
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
Optical Rotation +56° (Neat)
Boiling Point 195°C

Specifications

Chemical Name or Material (1S,2S)-(+)-2-Methoxycyclohexanol
CAS 134108-92-2
Molecular Formula C7H14O2
Formula Weight 130.19
MDL Number MFCD00211253
Beilstein 1848990
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.