Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Methoxycyclohexanol, ChiPros 99%, ee 98%


Quantity 5g

Manufacturer: alfa aesar™  L1906506

 View more versions of this product

Catalog No. AAL1906506

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 130.19
Molecular Formula C7H14O2
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material (1S,2S)-(+)-2-Methoxycyclohexanol
CAS 134108-92-2
Assay 99%
MDL Number MFCD00211253
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Beilstein 1848990
Health Hazard 1 Warning
Optical Rotation +56° (Neat)
Assay Percent Notes ee 98%
Boiling Point 195°C
Quantity 5g

Specifications

Chemical Name or Material (1S,2S)-(+)-2-Methoxycyclohexanol
CAS 134108-92-2
Molecular Formula C7H14O2
Formula Weight 130.19
MDL Number MFCD00211253
Beilstein 1848990
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents